Skip to Content
Merck
All Photos(1)

Documents

SAYB02

Supelco

Saybolt Colour Reference Standard

ASTM D 6045, D 156 (ASTM colour 25)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

according to ASTM® D 156
according to ASTM® D 6045

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

The Saybolt Colour Reference Standard has its spectral transmittance measured from 380 nm to 780 nm at intervals of 1 nm, using a Perkin-Elmer Lambda 650 spectrophotometer. The colour values provided for the reference standard are within the ASTM D6045 specification data table X1.3 colour range of Saybolt Colour 25 ± 1. It is manufactured, characterized, and distributed meeting the requirements of ISO/IEC 17025. Please note that the value stated on this page is nominal. All certified values for the current lots of all Saybolt Colour Reference Standards can be found on the Paragon Scientific webpage

Application

Saybolt Colour Reference Standard finds use in calibration of colour measuring instruments and validation of test results.

Features and Benefits

  • Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory under UKAS accreditation
  • Traceable to international standards
  • Available in 500 mL quantity with tamper evident packaging
  • 18 months shelf life
  • Accompanied with a latest EU compliant MSDS available in 24 languages

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F

Flash Point(C)

76 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service