Skip to Content
Merck
All Photos(1)

Key Documents

SAAR1260520

Barium chloride dihydrate

univAR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaCl2 · 2H2O
CAS Number:
Molecular Weight:
244.26
MDL number:
UNSPSC Code:
12352302
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

product line

univAR

packaging

poly bottle of 1 × 500 g (SAAR1260520EM)

storage temp.

2-30°C

SMILES string

O.O.Cl[Ba]Cl

InChI

1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

PWHCIQQGOQTFAE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service