Skip to Content
Merck
All Photos(1)

Key Documents

PHR9274

Supelco

2-Mercaptoethanol

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

PHR9274-100MG
1.460,00 kr.

1.460,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
PHR9274-100MG
1.460,00 kr.

About This Item

Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Beilstein:
773648
MDL number:
UNSPSC Code:
41116107

1.460,00 kr.


Please contact Customer Service for Availability

grade

certified reference material
TraceCERT®

Quality Level

Agency

Not traceable to any primary pharmacopeia standard

vapor density

2.69 (vs air)

vapor pressure

1 mmHg ( 20 °C)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

expl. lim.

18 %

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

14.3 M (pure liquid)

refractive index

n20/D 1.500 (lit.)

bp

157 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

storage temp.

−20°C

SMILES string

OCCS

InChI

1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2

InChI key

DGVVWUTYPXICAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 2 Oral

Target Organs

Liver,Heart

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service