Skip to Content
Merck
All Photos(1)

Documents

PHR9070

Supelco

Favipiravir

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

6-Fluoro-3-hydroxy-2-pyrazinecarboxamide, 6-fluoro-3,4-dihydro-3-oxo-2-pyrazinecarboxamide, Avigan, T-705, T705

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4FN3O2
CAS Number:
Molecular Weight:
157.10
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

pharmaceutical small molecule

storage temp.

−20°C

SMILES string

FC1=CNC(C(C(N)=O)=N1)=O

InChI

1S/C5H4FN3O2/c6-2-1-8-5(11)3(9-2)4(7)10/h1H,(H2,7,10)(H,8,11)

InChI key

ZCGNOVWYSGBHAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Biochem/physiol Actions

Favipiravir (T-705) is an inhibitor of viral RNA polymerase. In cells, the comound is converted to T-705-4-ribofuranosyl-5-triphosphate, which selectively targets the viral polymerase without affecting mammalian RNA or DNA polymerases. Favipiravir inhibits replication of inluenza viruse A, B and C with IC50 values between 0.013 and 0.48 mM. The compound protects mice from lethal-dose infections of wild type and neuraminidase inhibitor-resistant strains of H5N1 viruses, Favipiravir has been found to have activity against many viruses including flavi-, alpha-, filo-, bunya-, arena-, noro-, and of other RNA viruses, and is already used off-label for Ebola and Lassa virus.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service