Skip to Content
Merck
All Photos(1)

Documents

PHL82307

L-Arginine

phyproof® Reference Substance

Synonym(s):

(S)-2-Amino-5-guanidinopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
174.20
Beilstein:
1725413
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

mp

222 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1

InChI key

ODKSFYDXXFIFQN-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service