Skip to Content
Merck
All Photos(1)

Key Documents

K5163-U

Omnifit® EZ Benchmark Replacement Glass Columns

I.D. × L 15 mm × 330 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
1.250,00 kr.

1.250,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
1.250,00 kr.

About This Item

UNSPSC Code:
23151816
NACRES:
SB.52

1.250,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

manufacturer/tradename

Omnifit 006ERG-15-33

Quality Level

technique(s)

LPLC: suitable

I.D. × L

15 mm × 330 mm

Looking for similar products? Visit Product Comparison Guide

General description

Replacement glass columns are available in a wide range of bore sizes and lengths. This not only enables you to replace a column should breakage occur, but allows you to obtain several different length columns to use with endpieces you already have for that bore size. The modular nature of our column ranges means that for the ultimate flexibility you can mix and match column sizes with different endpiece types to your specific requirements.

Legal Information

Omnifit is a registered trademark of Diba Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service