Skip to Content
Merck
All Photos(1)

Key Documents

G005490

SFC Analysis of 1-(1-Naphthyl)ethylamine Enantiomers on Astec® Cellulose DMP

application for SFC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

test parameters

column: Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase: 20% methanol (0.1% MSA, methanesulfonic acid) in CO2
flow rate: 3 mL/min
pressure: 1740 psi (120 bar)
column temp.: 40 °C
detector: UV, 230 nm
injection: 5 μL
sample: 2 mg/mL in methanol

suitability

application for SFC

application(s)

chemicals and industrial polymers

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    1-(1-Naphthyl)ethylamine, 98%

mobile phase component

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service