Skip to Content
Merck
All Photos(1)

Documents

96957

Supelco

Diphenyl carbonate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)2CO
CAS Number:
Molecular Weight:
214.22
Beilstein:
1074863
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

technique(s)

HPLC: suitable

bp

301-302 °C (lit.)

mp

79-82 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

O=C(Oc1ccccc1)Oc2ccccc2

InChI

1S/C13H10O3/c14-13(15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H

InChI key

ROORDVPLFPIABK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

334.4 °F - closed cup

Flash Point(C)

168 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service