Skip to Content
Merck
All Photos(1)

Key Documents

822858

TSKgel® SuperSW mAb HTP (4 µm) HPLC Column

L x I.D. 2 cm × 3 mm, diol phase

Synonym(s):

TSKgel® Size Exclusion (SW-Type) HPLC Guard Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
4.830,00 kr.

4.830,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
4.830,00 kr.

About This Item

UNSPSC Code:
41115711

4.830,00 kr.


Please contact Customer Service for Availability

Product Name

TSKgel® SuperSW mAb HTP Guard Column, phase diol, L × I.D. 2 cm × 3 mm, 4 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh SuperSW mAb HTP Guard Column, 22858

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
0.5 mL/min flow rate
10-30 °C temperature

technique(s)

HPLC: suitable

L × I.D.

2 cm × 3 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

4 μm

pore size

250 Å pore size

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

The TSKgel SuperSW and UltraSW mAb columns consist of three specialized columns designed for the separation and analysis of monoclonal antibodies (mAb). While mAbs can be analyzed using many different modes of HPLC, size exclusion is best for aggregation, dimer, and fragmentation, making it the best method for heterogeneity studies.TSKgel SuperSW mAb HTP columns provide high throughput analysis of mAb monomer and dimer and is UHPLC compatible. These short, narrow bore (15 cm x 4.6 mm ID) silica-based stainless steel columns offer reduced lot-to-lot variation, long column life, reduction of unspecified adsorption, and superior recovery of aggregates. The TSKgel SW mAb HTP columns utilize a unique pore-controlled technology, which produces a shallow calibration curve in the molecular weight region of a typical monoclonal antibody.

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service