Skip to Content
Merck
All Photos(1)

Key Documents

821833

TOYOPEARL GigaCap® S-650M Bulk Media

bottle of 100 mL, phase sulphonyl, 75 μm particle size

Synonym(s):

TOYOPEARL GigaCap® S-650 Cation Exchange Media

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
4.060,00 kr.

4.060,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 ML
4.060,00 kr.

About This Item

UNSPSC Code:
41115700

4.060,00 kr.


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 100 mL

manufacturer/tradename

Tosoh 21833

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

sulphonyl phase

particle size

75 μm

operating pH

3-13

capacity

156 g/L±20 adsorption capacity (hIgG)

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl GigaCap S-650 resin is a high capacity, high resolution cation exchange resin* optimized for process scale applications, including the capture and purification of IgG as well as the polishing of monoclonal antibodies. A Toyopearl HW-65 polymeric bead has been chemically modified to provide a higher number of cationic binding sites for increased resin capacity.
While Toyopearl GigaCap S-650M resin was specifically optimized for IgG, with dynamic binding capacities in the vicinity of 140 mg/mL for IgG, it also exhibits high binding capacities for smaller proteins. Typical dynamic binding capacities for lysozyme are in excess of 160 mg/mL.
Toyopearl GigaCap S-650S resin has dynamic binding capacities approaching 165 g/L for gamma globulin. This high capacity, combined with the 35 μm mean particle size, makes Toyopearl GigaCap S-650S ideal for high resolution applications.
Toyopearl GigaCap S-650 resin is stable at high linear velocities, has good pressure-flow characteristics and will withstand back pressures up to 3 bar. The higher capacity and back pressure stability of Toyopearl GigaCap S-650 resin creates opportunities for increased throughput in downstream cation exchange purification steps.
Toyopearl GigaCap S-650M resin also has excellent base stability in 1.0 mol/L NaOH and shows robust clean in place (CIP) properties with minimal loss of capacity after 50 X 1 hour cycles of caustic exposure also using 1.0 mol/L NaOH. (data)
*specifically designed for packed bed use

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation
Toyopearl GigaCap is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service