Skip to Content
Merck
All Photos(1)

Key Documents

82031-U

Hamilton® HC-75 (9 µm) HPLC Columns

L × I.D. 25 cm × 4.1 mm, stainless steel hardware

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

Product Name

Hamilton HC-75 HPLC Column, 9 μm particle size, L × I.D. 25 cm × 4.1 mm, stainless steel hardware

material

stainless steel column

Agency

suitable for USP L19

manufacturer/tradename

Hamilton

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.1 mm

matrix

polymeric

particle size

9 μm

operating pH

1-14

separation technique

ion exclusion

Looking for similar products? Visit Product Comparison Guide

General description

Hamilton HC-75 columns are used for the separation of mono and disaccharides, orgainc acids, sugars, and sugar alcohols.

Footnote

Ca2+ Form

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service