Skip to Content
Merck
All Photos(1)

Key Documents

817088

TSKgel® DNA-NPR (5 µm) HPLC Columns

L x I.D. 0.5 cm × 4.6 mm, DEAE (diethylaminoethyl) phase

Synonym(s):

TSKgel® DEAE-NPR HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
4.200,00 kr.

4.200,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
4.200,00 kr.

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

4.200,00 kr.


Please contact Customer Service for Availability

Product Name

TSKgel® DNA-NPR Guard Column, phase DEAE (diethylaminoethyl), L × I.D. 0.5 cm × 4.6 mm, 5 μm particle size

material

PEEK

product line

TSKgel®

feature

counter ion Cl-

manufacturer/tradename

Tosoh 17088

parameter

≤1 M salt concentration
≤20% organic solvent
1.6 mL/min flow rate
10-60 °C temperature

technique(s)

HPLC: suitable

L × I.D.

0.5 cm × 4.6 mm

matrix

silica particle platform

matrix active group

DEAE (diethylaminoethyl) phase

particle size

5 μm

operating pH

2-12

separation technique

anion exchange

General description

Guard Column for 4.6mmID TSKgel DEAE-NPR column

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service