Skip to Content
Merck
All Photos(1)

Key Documents

807997

TOYOPEARL® SP-650M Bulk Media

phase sulfopropyl, bottle of 250 mL, 65 μm particle size

Synonym(s):

TOYOPEARL® SP-650 Cation Exchange Media

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
4.220,00 kr.

4.220,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
250 ML
4.220,00 kr.

About This Item

UNSPSC Code:
41115700

4.220,00 kr.


Please contact Customer Service for Availability

description

SP-650M

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 250 mL

parameter

3 bar max. pressure

technique(s)

HPLC: suitable

matrix

methacrylate

matrix active group

sulfopropyl phase

particle size

65 μm

operating pH

3-11

capacity

0.15 meq/mL±0.02 meq/mL ion exchange capacity
50 g/L±10 adsorption capacity (lysozyme)
55 mg/mL±10 mg/mL binding capacity (lysozyme)

compatibility

mode of use strong cation exchange chromatography

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl SP-650 resin is a strong cation exchanger for biomolecule purification. It can be used for high throughput capture, intermediate purification, and polishing process steps. If more powerful resolution is needed the smaller particle TSKgel SP-5PW resin should be used. Selectivity remains the same.

Application

TOYOPEARL® SP-650M columns are used in separation media and ion exchange resins. TOYOPEARL® SP has been used in studies contributing to the improved healing of wounds on the skin, cornea and gastrointestinal tract.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Sharma et al.
Protein expression and purification, 60(1), 7-14 (2008-04-24)
Amongst the various endogenous growth factors, epidermal growth factor (EGF) plays an important role in normal wound healing of tissue such as skin, cornea and gastrointestinal tract. Various studies have proved that supplementing recombinant human EGF (rhEGF) results in significant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service