Skip to Content
Merck
All Photos(1)

Key Documents

806729

TSKgel® G4000SW (17 µm) HPLC Columns

L x I.D. 30 cm × 21.5 mm, diol phase

Synonym(s):

TSKgel® Size Exclusion (SW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
46.230,00 kr.

46.230,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
46.230,00 kr.

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

46.230,00 kr.


Please contact Customer Service for Availability

Product Name

TSKgel® G4000SW HPLC Column, phase diol, L × I.D. 30 cm × 21.5 mm, 17 μm particle size

material

stainless steel hardware

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 06729

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
10 bar max. pressure
10-30 °C temperature
8 mL/min flow rate

technique(s)

HPLC: suitable

L × I.D.

30 cm × 21.5 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

17 μm

pore size

450 Å

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

As the first generation of silicia based SEC columns, the TSKgel G4000SW columns provide excellent separation of proteins and other biomolecules. TSKgel G4000SW columns are available in 13μm particle size in Glass and Stainless Steel analytical columns. Particle sizes of 13 and 17μm are used to pack 30cm and 60cm x 21.5mm ID columns, respectively, for semi-preparative purifications.

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service