79994
Trinexapac-ethyl
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
format
neat
storage temp.
2-8°C
SMILES string
O=C(CC(C(OCC)=O)CC1=O)C1=C(C2CC2)O
InChI
1S/C13H16O5/c1-2-18-13(17)8-5-9(14)11(10(15)6-8)12(16)7-3-4-7/h7-8,16H,2-6H2,1H3/b12-11-
InChI key
RVKCCVTVZORVGD-QXMHVHEDSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Trinexapac-ethyl (TE) is a plant growth regulator belonging to the cyclohexanones group commonly used in cereal production systems. It retards plant growth by inhibiting gibberellin (GA) biosynthesis, thereby reducing the risk of stem lodging.
As per Regulation (EC) No 1107/2009, repealing the directive 91/414/EEC, trinexapac (aka cimetacarb ethyl) is approved for use in the European Union. A maximum residue limit (MRL) of 0.01 mg/kg is applicable for the presence of trinexapac (sum of trinexapac (acid) and its salts, expressed as trinexapac) in fresh/frozen fruits, and tree nuts, in accordance with Regulation (EU) 2017/1016.
Application
The trinexapac-ethyl CRM may also find the following uses:
- To study the effect of growth regulator trinexapac-ethyl on oats yield in Brazil
- Determine the effects of TE dose and application timings on lodging, canopy architecture, grain yield, and quality of winter cereals
- To study the seed yield responses of annual ryegrass (Lolium multiflorum Lam.) to trinexapac-ethyl
- To determine the effects of TE application on Kentucky bluegrass (Poa pratensis L.) responses to salinity stress by greenhouse sand culture experiment
- To study the effects of trinexapac-ethyl on diploid red clover (Trifolium pratense L.) seed production in diverse environments
- To evaluate the effect of trinexapac-ethyl at two application times, before planting or after planting, in Eucalyptus urophylla under conditions of nutritional deficiency
Recommended products
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 1 - Skin Sens. 1B - STOT RE 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service