Skip to Content
Merck
All Photos(1)

Key Documents

77988

Supelco

Acetoacetic acid-13C4 lithium salt

analytical standard

Synonym(s):

Lithium acetoacetate-13C4

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
521,00 kr.
10 MG
1.120,00 kr.

521,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
521,00 kr.
10 MG
1.120,00 kr.

About This Item

Empirical Formula (Hill Notation):
13C4H6O3 · xLi+
Molecular Weight:
106.06 (free acid basis)
UNSPSC Code:
12352106
NACRES:
NA.24

521,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

SMILES string

[13CH3][13C]([13CH2][13C](O)=O)=O

InChI

1S/C6H10O4/c1-6(2,3-7)4(8)5(9)10/h7H,3H2,1-2H3,(H,9,10)

InChI key

PKVVTUWHANFMQC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service