Skip to Content
Merck
All Photos(1)

Key Documents

67109

Supelco

Potassium hydroxide solution

4 M, eluent concentrate for IC

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
749,00 kr.

749,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
749,00 kr.

About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352320
NACRES:
NA.21

749,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

concentration

4 M in H2O
4 M

SMILES string

[K+].[O-][H]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the Fluoride, Chloride, Sulfate anions specification for this 4M KOH solution?

    1 answer
    1. The trace analysis of this product does not include Fluoride. The maximum amount allowable for Chloride and Sulfate are 40 mg/Kg and 20 mg/Kg, respectively. These values are reported on a lot-by-lot basis. Please see the link below to access a sample of lot-specific Certificate of Analysis:
      https://www.sigmaaldrich.com/product/supelco/67109#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service