Skip to Content
Merck
All Photos(1)

Documents

575664-U

Supelco

Supel-Select SCX SPE 96-well Plate

bed wt. 30 mg/well, pk of 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104018
NACRES:
NB.21

material

PE frit (20 μm)
polypropylene hardware

quality

MS Friendly

composition

bed wt., 30 mg/well

packaging

pk of 1

technique(s)

solid phase extraction (SPE): suitable

surface area

160-420 m2/g

matrix

Sulfonic acid functionalized hydrophilic modified styrene polymer particle platform

matrix active group

sulfonic acid phase

particle size

50-70 μm

pore size

80-200 Å pore size

application(s)

food and beverages

separation technique

ion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Ion-Exchange
Sample Matrix Compatibility: Aqueous solutions (biological fluids, water)

  • Sulfonic acid functionalized hydrophilic modified styrene polymer
  • Developed for the extraction of a highly broad range of compounds from aqueous samples
  • Retention predominately based on ion-exchange; however, because the phase is hydrophilic modified, it is amenable to polar compounds as well.

Features and Benefits

  • Excellent sample prep performance at a lower price
  • Amenable to generic methodology - save time, money, and headache during method development
  • Greater capacity allows for smaller bed weights = smaller elution volumes = time savings in sample processing
  • Resistance to overdrying allows for more robust methdology
  • Low UV and MS extractables for lower background and greater sensitivity
  • Stringent production and QC guidelines offer greater lot-to-lot, tube-to-tube, and well-to-well consistency for improved accuracy and precision

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

SPE retention mechanism in this case is based on the electrostatic attraction of charged functional groups of the analyte(s) to oppositely charged functional groups on the sorbent.

Protocols

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

In this study, optimized methods are presented for sample preparation and chiral chromatography for the LC/MS analysis of amphetamine and methamphetamine enantiomers in urine.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service