55522-U
Supel™Genie RP-Amide Online SPE Cartridges
L × I.D. 20 mm × 4.0 mm, pkg of 6 ea
About This Item
material
stainless steel tube
product line
Supel™
composition
bed wt., 180 mg
packaging
pkg of 6 ea
technique(s)
solid phase extraction (SPE): suitable
L × I.D.
20 mm × 4.0 mm
matrix active group
RP-Amide
application(s)
food and beverages
Looking for similar products? Visit Product Comparison Guide
General description
Supel Genie Online SPE cartridges are designed for direct injection of untreated samples (e.g. water and urine) or processed samples (e.g. protein-precipitated plasma and serum), for analysis with liquid chromatography and mass spectrometry (LC/MS). The Supel Genie Online SPE cartridges enable online sample cleanup, concentration, and separation with excellent reproducibility and performance. Coupled with mass spectrometry for detection, the Supel Genie Online SPE cartridges provide a simple, automated method for sample analysis and are ideal for use in the LC/MS/MS analysis of biological fluids such as plasma and serum.
For first time use, a starter kit is required to obtain reusable universal hardware that will fit all Supel Genie Online SPE cartridges (P/N 55516-U).
LC instrumentation must be configured to accommodate Online SPE cartridges. Please see instrument setup diagram in datasheet. 55522-U
Please contact [email protected] for more information on setup and hardware.
Application
Features and Benefits
- Provides cleaner samples for longer column life, less instrument downtime, and more accurate and reproducible data
- Automation saves time, decreases costs, and eliminates human error
- Highly reproducible, consistent product performance
- RP-Amide cartridges are ideal for reversed-phase extraction of nonpolar to polar compounds
- RP-Amide cartridges often offer improved performance for polar analytes, especially those that can interact via hydrogen bonding, when compared to standard alkyl phases
- Cartridges also available in pack of 2
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service