Skip to Content
Merck
All Photos(1)

Documents

51177-U

OPTI-GUARD® C18 (40 μm) HPLC Guard Column

1mm HPLC Guard Column, pkg of 5 ea

Synonym(s):

High-Performance Guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

OPTI-GUARD® 1mm Guard Column, C18 (violet label), pkg of 5 ea

packaging

pkg of 5 ea

manufacturer/tradename

Opti-Guard®

matrix active group

C18 (octadecyl) phase

particle size

40 μm

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

OPTI-GUARD® sets the standard for low-impact, easy to use pre-column protection. Designed for use with analytical (4.6 mm, 3.0 mm I.D.) and narrow bore (2.1 mm, 1.0 mm) columns, the patented floating stem design automatically adjusts to any manufactures tube stop depth for a zero-dead volume connection every time. The OPTI-GUARD® 1 mm requires no special connecting hardware or tools for installation.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

OPTI-GUARD is a registered trademark of Optimize Technologies, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service