Skip to Content
Merck
All Photos(1)

Key Documents

504254

Supelco

Supelguard HPLC Guard Cartridge Holder

Direct-Connect (Swivel-type), for use with Supelguard cartridges (2 cm L. x 3 to 4.0mm I.D.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

Product Name

Supelguard Guard Cartridge Holder, Direct-Connect (Swivel-type), for use with Supelguard cartridges (2 cm L. x 3 to 4.0mm I.D.)

material

stainless steel holder

description

swivel design

packaging

pkg of 1 ea

fitting

10-32 female UNF (Valco compatible)
5/16-40 female UNF (threads directly to column cartridge)

compatibility

for use with Supelguard cartridges (2 cm L. x 3 to 4.0mm I.D.)

General description

Supelguard Guard Cartridge is a stand-alone holder for 2 cm Guard Column. It holds 2.1 mm, 3.0 mm, or 4.0 mm ID Supelguard cartridge. It includes 1/16″ nuts and ferrules and 2″/5 cm of 0.01″ ID x 1/16″ OD tubing. As a direct-connect Guard Column Holder, it directly connects a 2 cm Supelguard cartridge to a Supelco analytical cartridge after the column end-fitting is removed.

Footnote

Integral guard column holder fits 2 cm x 2, 3, and 4 mm I.D. Supelco and Astec® HPLC columns. It couples directly to the HPLC column, and uses standard 1/16" O.D. Valco-type nuts and ferrules on the freely-rotating inlet port. Cartridges, tubing, nuts, and ferrules are not included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service