Skip to Content
Merck
All Photos(1)

Documents

503231

Supelco

Mounting Clip

for use with 200 cc traps

Synonym(s):

CLIP FOR 200CC MOLECULAR SIEVE TRAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
eCl@ss:
32110204

packaging

pkg of 1 ea

compatibility

for use with 200 cc traps

Looking for similar products? Visit Product Comparison Guide

General description

This clip is designed to fit one of our in-line traps based on the volume of the trap. Simply mount two or more clips to a bench or wall using screws (not supplied), then snap the trap into the clips. One plastic clip per package.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service