Key Documents
502383
Micro-Splitter Valve (Low Pressure)
1/4-28 flangeless fittings
Select a Size
1 EA
812,00 kr.
812,00 kr.
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
812,00 kr.
About This Item
UNSPSC Code:
41115854
eCl@ss:
32060101
812,00 kr.
Please contact Customer Service for Availability
Request a Bulk Order
feature
graduated
manufacturer/tradename
Upchurch Scientific, Inc P-450
parameter
800 psi max. pressure
fitting
1/4-28 flangeless fittings
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)