Skip to Content
Merck
All Photos(1)

Key Documents

442483

Supelco

Benzylamine

analytical standard

Synonym(s):

α-Aminotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
741984
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.543 (lit.)

bp

184-185 °C (lit.)

mp

10 °C (lit.)

solubility

H2O: soluble

density

0.981 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

NCc1ccccc1

InChI

1S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5H,6,8H2

InChI key

WGQKYBSKWIADBV-UHFFFAOYSA-N

Gene Information

human ... AOC3(8639)
mouse ... Aoc3(11754)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saioa Gomez-Zorita et al.
Journal of physiology and biochemistry, 69(3), 585-593 (2013-01-15)
Resveratrol is a naturally occurring polyphenol found in many dietary sources and red wine. Recognized as a cancer chemoprevention agent, an anti-inflammatory factor and an antioxidant molecule, resveratrol has been proposed as a potential anti-obesity compound and to be beneficial
Zi Jun Wang et al.
ChemSusChem, 8(20), 3459-3464 (2015-09-10)
Metal-free visible-light photocatalysts offer a clean, sustainable solution to many pressing environmental issues. Herein, we present a molecular design strategy to fine-tune the valence and conduction band levels of a series of conjugated microporous polymer networks based on poly(benzochalcogenadiazole) for
Danielle M Shacklady-McAtee et al.
Organic letters, 13(13), 3490-3493 (2011-06-08)
A nickel(0) catalyst effectively mediates the cyclization of N-benzoyl aminals in the presence of a stoichiometric Lewis acid. This method enables preparation of a variety of isoindolinones with substitution on the benzoyl fragment and C-3 carbon. This reaction likely proceeds
Farid Oukacine et al.
Analytical chemistry, 84(7), 3302-3310 (2012-03-28)
A new methodology for an antibacterial assay based on capillary electrophoresis with multiple UV detection points has been proposed. The possible antibacterial activity of cationic molecules on bacteria (Gram-positive and Gram-negative) is studied by detecting the bacteria before, during, and
Fei Mao et al.
Bioorganic & medicinal chemistry, 20(19), 5884-5892 (2012-09-05)
In an effort to identify novel multifunctional drug candidates for the treatment of Alzheimer's disease (AD), a series of hybrid molecules were synthesised by reacting N-(aminoalkyl)tacrine with salicylic aldehyde or derivatives of 2-aminobenzaldehyde. These compounds were then evaluated as multifunctional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service