Skip to Content
Merck
All Photos(1)

Key Documents

40567

Supelco

7,12-Dimethylbenz[a]anthracene solution

certified reference material, 1000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1912135
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

Quality Level

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Cc1c2ccccc2c(C)c3c1ccc4ccccc34

InChI

1S/C20H16/c1-13-16-8-5-6-9-17(16)14(2)20-18(13)12-11-15-7-3-4-10-19(15)20/h3-12H,1-2H3

InChI key

ARSRBNBHOADGJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7,12-Dimethylbenz[a]anthracene is a polyaromatic hydrocarbon, which is a pollutant that can be released into the environment due to incomplete combustion.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - Methanol

Flash Point(C)

9.7 °C - Methanol


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioprotective Efficacy of Erucin Against 7, 12-Dimethylbenz (?) anthracene-Induced Microstructural Changes in Male Wistar Rats
Mannan R, et al.
Turkish Journal of Pathology, 33, 150-156 (2017)
Ramar Perumal Samy et al.
PloS one, 7(12), e48514-e48514 (2013-01-04)
Breast cancer is the most common cancer among women. To date, improvements in hormonal and cytotoxic therapies have not yet led to a sustained remission or cure. In the present study, we investigated the in vitro and in vivo antitumor
Peter T Vedell et al.
Molecular pharmacology, 83(3), 698-708 (2013-01-08)
Examination of three retinoid X receptor (RXR) agonists [Targretin (TRG), UAB30, and 4-methyl-UAB30 (4-Me-UAB30)] showed that all inhibited mammary cancer in rodents and two (TRG and 4-Me-UAB30) strikingly increased serum triglyceride levels. Agents were administered in diets to female Sprague-Dawley
Gaurav Kumar et al.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer, 31(4), 295-312 (2013-02-12)
In the present study, post-treatment effects of dietary turmeric on markers related to apoptosis, cell proliferation, and inflammation in 7,12-dimethylbenz(a)anthracene (DMBA)-induced hamster buccal pouch (HBP) tumors were investigated. Tumors were induced by applying 0.5% DMBA topically to the HBP three
S Manoharan et al.
Asian Pacific journal of cancer prevention : APJCP, 13(11), 5701-5708 (2013-01-16)
The aim of the study was to investigate the chemopreventive potential of andrographolide in 7,12-dimethylbenz(a) anthracene (DMBA)-induced hamster buccal pouch carcinogenesis. Oral tumors developed in the buccal pouch of golden Syrian hamsters at a 100% incidence on painting with 0.5%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service