Skip to Content
Merck
All Photos(1)

Key Documents

27235-U

Supelco

Stoppers

stopper diam. 20 mm, silicone, pkg of 100 ea

Synonym(s):

Rubber Stoppers

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
965,00 kr.

965,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 EA
965,00 kr.

About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

965,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

material

silicone

packaging

pkg of 100 ea

O.D.

20 mm

stopper diam.

20 mm

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the Durometer reading and color of the stoppers 27235-U?

    1 answer
    1. The product is a red Silicone product with a 30 durometer.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service