Skip to Content
Merck
All Photos(1)

Key Documents

24643

Supelco

Battery charger

for use with Model 1060 Bag Sampler, AC/DC input 110 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

technique(s)

whole air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Model 1060 Bag Sampler

Looking for similar products? Visit Product Comparison Guide

General description

Battery charger is a 110V AC charger used as accessories for field sampling pumps. It is used with a DC battery in a 1-liter vacuum bag sampler.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service