Key Documents
22434
Mirror with Rotating Head
chrome-plated handle, pkg of 1 ea
About This Item
UNSPSC Code:
41100000
material
chrome-plated handle
packaging
pkg of 1 ea
General description
Ideal for examining inside of injection and detector fittings for ferrule or column fragments. Rotating head allows wide range of movement.
Choose from one of the most recent versions:
Certificates of Analysis (COA)