Key Documents
20392
In-Line Pressure Gauge Kit
About This Item
UNSPSC Code:
41112500
parameter
0-100 psi pressure range
technique(s)
gas chromatography (GC): suitable
General description
Install a pressure gauge in the gas delivery system to measure pressure at any given point. For example, install one upstream and downstream of the High Capacity Gas Purifier. When the pressure drop across the purifier exceeds 10 psi, the converter tube should be replaced. Kit contains:
  • 2 in. (5 cm) diameter steel and copper alloy gauge (0-100 psi)
  • NPT to Swagelok adapter
  • 1/8 in. tee
  • 18 in. (1/2 m) of 1/8 in. copper line
  • assembly and installation instructions
Nuts and ferrules included.
Application
In-line pressure gauge kit may be used to monitor the column inlet pressure of high-performance liquid chromatography (HPLC) equipment, which is further modified for carbon-dioxide supercritical fluid chromatography (SFC).[1]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Modification of conventional high-performance liquid chromatography equipment for use in carbon dioxide supercritical fluid chromatography.
Simpson CR, et al.
Journal of Chromatography A, 371, 109-119 (1986)