Skip to Content
Merck
All Photos(6)

Key Documents

ZRB2997

Sigma-Aldrich

Anti-c-Rel Antibody, clone 4G9 ZooMAb® Rabbit Monoclonal

enhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

Proto-oncogene c-Rel, REL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4G9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 68.52 kDa
observed mol wt ~79 kDa

purified by

using Protein A

species reactivity

rat, mouse, human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... REL(5966)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4G9 is a ZooMAb® rabbit recombinant monoclonal antibody that detects Proto-oncogene c-Rel. It targets an epitope within 22 amino acids from the C-terminal region of human c-Rel.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal region t of Human c-Rel.

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Proto-oncogene c-Rel in Jurkat cell lysate.

Tested Applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected c-Rel in Daudi, Jurkat, and MCF-7 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected c-Rel in Rat spleen tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Proto-oncogene c-Rel (UniProt: Q04864; also known as c-Rel, REL) is encoded by the REL gene (Gene ID: 5966) in human. c-Rel is one of the five subunits of the Nuclear Factor kappa B (NF- B) family of proteins. It can act as a transcriptional activator by binding to specific DNA sequences ( B sites). It contains several conserved domains, including an N-terminal DNA-binding domain, a dimerization domain called the Rel homology domain, and a C-terminal transcriptional activation domain that enables c-Rel to bind to DNA and regulate gene expression. c-Rel forms a dimer mostly with the p50 or RelA NF- B subunits, but other combinations can also occur. It is a cellular homologue of the avian Rev-T retroviral oncoprotein v-Rel. It plays a crucial role in immune and inflammatory responses, regulating B- and T-cell proliferation and apoptosis. In addition, it may play a role in tumorigenesis, cell growth and differentiation. c-Rel expression can be regulated by various signaling pathways and external stimuli, such as cytokines and microbial components. It is expressed in most cells, with high expression in B- and T-cells. Mutations and aberrant activation of c-Rel has been associated with several diseases, including cancer, autoimmune disorders, and inflammatory conditions. In cancer, c-Rel can promote fibrosis, tumor growth, invasion, and metastasis by regulating genes involved in cell proliferation, angiogenesis, and apoptosis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Basavarajappa, SC. and Ramakrishnan, P. (2020) Cell. Mol. Life Sci., 77(17); 3325-3340).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service