Skip to Content
Merck
All Photos(6)

Key Documents

ZRB2787

Sigma-Aldrich

Anti-SLC31A1/CTR1 Antibody, clone 1K21 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Copper transporter 1, High affinity copper uptake protein 1, Solute carrier family 31 member 1, hCTR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1K21
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 21.09 kDa
observed mol wt ~28 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... SLC31A1(1317)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1K21 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects High affinity copper uptake protein 1 (SLC31A1). It targets an epitope within 18 amino acids from the N-terminal extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal, extracellular domain of human High affinity copper uptake protein 1 (SLC31A1).

Application

Quality Control Testing

Evaluated by Western Blotting in Human liver cell membrane preparations.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SLC31A1/CTR1 in Human liver cell membrane preparations.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SLC31A1/CTR1 in Jurkat cell membrane preparations.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SLC31A1/CTR1 in HepG2 cells.

Affinity Binding Assay: A representative lot of this antibody bound SLC31A1/CTR1 peptide with a KD of 1.0 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

High affinity copper uptake protein 1 (UniProt: O15431; also known as Copper transporter 1, hCTR1, Solute carrier family 31 member 1) is encoded by the SLC31A1 (also known as COPT1, CTR1) gene (Gene ID: 1317) in human. SLC31A1 is a homotrimeric, multi-pass membrane protein with two extracellular domains (aa 1-61; 154-156), three transmembrane domains (aa 62-82; 133-153; 157-177), and two cytoplasmic domains (aa 83-132; 178-190). It serves as a high-affinity copper uptake protein that transports copper1+ from extracellular space to the cytoplasm and delivers to specific chaperones, such as Antioxidant 1 Copper Chaperone (ATOX1). The copper1+ transport mechanism is reported to be sodium-independent and is saturable. This transport is stimulated by extracellular acidic pH and high levels of potassium ions. Copper1+ import is reported to be regulated by a copper1+-dependent recycling of SLC31A1. It also participates in the first step of copper2+ acquisition by cells through a direct transfer of copper2+ from copper2+ carriers in blood then leading to copper2+ reduction and stabilization of copper1+. Its C-terminal domain is known to mediate copper1+ binding and is involved in the copper1+-dependent-ATOX1 interaction. It also limits transport through the pore by regulating the rate of exit of copper ions. Its N-terminal domain can collect copper2+ from copper2+ carriers in blood. SLC21A1 interacts with SLC31A2, and this interaction is shown to stabilize SLC31A2 and protect it from ubiquitination and degradation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Stefaniak, E., et al. (2018). Metallomics. 10(12); 1723-1727; Maryon, EB., et al. (2013). J. Biol. Chem. 288(25); 18035-18046; Lee, J., et al. (2002). J. Biol. Chem. 277(6); 4380-4387).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service