Skip to Content
Merck
All Photos(9)

Documents

ZRB2231

Sigma-Aldrich

Anti-LAG3 Antibody, clone 1B14-H1 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CD223, Lymphocyte activation gene 3 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B14-H1, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... LAG3(3902)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B14-H1 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects LAG3. It targets an epitope within 16 amino acids from the extracellular domain from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the extracellular domain within the N-terminal half of human LAG3.

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LAG3 in Jurkat cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LAG3 in Human tonsil tissue lysates and recombinant Mouse LAG3.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected LAG3 in one million Human peripheral blood mononuclear cells (PBMC).

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected LAG3 in Human tonsil tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound LAG3 peptide with a KD of 1.0 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Lymphocyte activation gene 3 protein (UniProt: P18627; also known as LAG-3, CD223) is encoded by the LAG3 (also known as FDC) gene (Gene ID: 3902) in human. LAG3 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 23-450), a transmembrane domain (aa 451-471), and a cytoplasmic domain (aa 472-525). It contains three Ig-like C2-type domains and one Ig-like V-type domain. LAG3 is primarily expressed in activated T-cells and in a subset of natural killer cells. Its expression is induced by IL-2, IL-7, and IL-12 on activated T-cells. Almost all CD4+ cells expressing LAG3 also Express CD25. However, only about 10% of CD4+ cells lacking LAG3 Express CD25. LAG3 serves as an inhibitory receptor on antigen activated T-cells and delivers inhibitory signals upon binding to ligands. Fibrinogen-like protein 1 (FGL1) is shown to be a major ligand for LAG3. LAG3 is reported to suppresses homeostasis proliferation (HP®) of CD4 and CD8 lymphocytes and some dendritic cell populations. Mice deficient in LAG3 display enhanced homeostasis proliferation (HP®). Following TCR engagement, it is shown to associate with CD3-TCR in the immunological synapse and directly inhibits T-cell activation. LAG3 can be proteolytically cleaved by ADAM10 and ADAM17 within the connecting peptide region to generate a secretory form (sLAG3). ADAM10 is reported to mediate constitutive cleavage, but cleavage increases following T-cell activation. On the other hand, cleavage by ADAM17 is induced by TCR signaling in a Protein Kinase C theta (PRKCQ)-dependent manner. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Scurr, M., et al. (2014). Mucosal Immunol. 7(2); 428-439; Goldberg, MV., and Drake, CG (2011). Curr. Top. Microbiol. Immunol. 344; 269-278).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
Express is a trademark of Merck KGaA, Darmstadt, Germany
HP is a registered trademark of Hewlett-Packard Corp.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service