Skip to Content
Merck
All Photos(21)

Documents

ZRB1705

Sigma-Aldrich

Anti-CTGF Antibody, clone 1O16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CCN family member 2;Cellular communication network factor 2;Connective tissue growth factor;Hypertrophic chondrocyte-specific protein 24;Insulin-like growth factor-binding protein 8;IBP-8;IGF-binding protein 8;IGFBP-8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O16, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 38.09 kDa
observed mol wt ~36 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CCN2(1490)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1O16 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Connective tissue growth factor (CTGF). It targets an epitope within 21 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human Connective tissue growth factor (CTGF).

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected CTGF in human kidney tissue sections.

Tested applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CTGF in human thymus tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound CTGF with a KD of 4.6 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CTGF in HeLa, , A431, HepG2, and NIH 3T3 cells.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected recombinant human CTGF.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

CCN family member 2 (UniProt: P29279; also known as Cellular communication network factor 2, Connective tissue growth factor, Hypertrophic chondrocyte-specific protein 24, Insulin-like growth factor-binding protein 8, IBP-8, IGF-binding protein 8, IGFBP-8) is encoded by the CCN2 (also known as CTGF, HCS24, IGFBP8) gene (Gene ID: 1490) in human. CTGF is a cysteine rich extracellular matrix monomeric protein of the CCN family that is synthesized with a signal peptide (aa 1-26), which is subsequently cleaved off to produce the mature form. It is a key signaling and regulatory molecule involved in different biological processes, including cell proliferation, angiogenesis, and wound healing, and in pathological events, such as tumor development and tissue fibrosis. It also serves as a major connective tissue mitoattractant secreted by vascular endothelial cells that promotes proliferation and differentiation of chondrocytes. Its expression is also observed in bone marrow and thymic cells. Four different domains of CTGF have been described: the insulin-like growth factor binding protein (IGFBP; aa 27-98), von Willebrand factor type C repeat (VWC; aa 101-167), thrombospondin type-1 repeat (TSP1; aa 198-243), and cysteine knot-containing domain (CT; aa 256-330). IGFBP and VWC domains constitute the N-terminal half of CTGF which is separated from the C-terminal half that contains TSP1 and CT domains by a hinge region. Due to the high number of disulfide bridges, most of the CTGF domains are resistant to proteolytic digestion, and most proteases primarily cleave only in the cysteine-free hinge region, producing almost exclusively intact N- and C-terminal half fragments. The truncated CTGF domains (N-terminal fragment and C-terminal fragment), can function independently to stimulate differentiation or proliferation of fibroblast and to increase collagen synthesis. The proper expression of CTGF is essential for physiological processes in of multiple organs. CTGF knockout mice display developmental skeletal malformations, and its high expression is reported to negatively regulate myelination during development and cause a range of neurodevelopmental disorders. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Chen, Z., et al. (2020). Front. Cell Dev. Biol. 8; 593269; Tsai, CC., et al. (2018). Sci. Rep. 8; Article 7276).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service