Key Documents
Z742618
Tube-O-Array dialyzer tray for Tube-O-DIALYZER
Select a Size
1 EA
1.760,00 kr.
1.760,00 kr.
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
1.760,00 kr.
About This Item
UNSPSC Code:
41123100
NACRES:
NB.24
1.760,00 kr.
Please contact Customer Service for Availability
manufacturer/tradename
G-BioSciences 786-145A
packaging
1 ea of
General description
Tube-O-Array Dialyzer is specifically developed for dialysis-equilibration of samples prior to 2D-gel analysis or other applications. Optimize up to 12 samples at a time. It consists of an Array Dialyzer Tray for holding 12 Tube-O-DIALYZER assemblies for simple and convenient equilibration?dialysis of the samples. It is suitable for 20μl to 2.5ml samples.
Legal Information
Tube-O-Array is a trademark of Geno Technology, Inc.
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)