Skip to Content
Merck
All Photos(1)

Key Documents

Z742004

Low Profile Roller Replacement O-Rings

Sign Into View Organizational & Contract Pricing

Select a Size

30 EA
434,00 kr.

434,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
30 EA
434,00 kr.

About This Item

UNSPSC Code:
41121815
NACRES:
NB.22

434,00 kr.


Please contact Customer Service for Availability

feature

fits 10ml - 50ml tubes

packaging

pkg of 30 ea

manufacturer/tradename

IBI ACCRLRN30

General description

There are 10 each for 10ml tubes, 10 each for 15ml tubes and 10 each for 50ml tubes. The O-rings slide onto the tubes and level them for rolling on the IBI Low Profile Roller.

A pack of tube O-rings is included with each new Low Profile Roller purchased.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service