Key Documents
Z693723
Duran® filtering flask, with Keck® assembly set
capacity 2000 mL
Synonym(s):
212046351
Select a Size
Z693723-1EA
1.040,00 kr.
1.040,00 kr.
Please contact Customer Service for Availability
Select a Size
Change View
Z693723-1EA
1.040,00 kr.
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
1.040,00 kr.
Please contact Customer Service for Availability
material
borosilicate glass
flat bottom flask
Joint
joint ST/NS neck60 mm I.D.
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 212046351
availability
not available in USA and Canada
capacity
2000 mL
General description
The Duran filtering flask includes a Keck clip assembly and is idea for vacuum filtrations. The plastic hose connections meet safety and product regulations, and the flask has the Erlenmeyer shape.
Manufactured from Duran borosilicate glass type 3.3, the flasks have maximum chemical and thermal resistance and meet ISO 6556 standard.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)