Z653683
Pyrex® glass culture tubes
O.D. × L 26 mm × 200 mm, wall thickness 2.0 mm, thread size 25 mm , cap
Synonym(s):
1636/44MP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass tube
round bottom
screw top cap
screw top neck
transparent tube
sterility
non-sterile
feature
neck thread external thread
packaging
pack of 40 ea
manufacturer/tradename
SciLabware 1636/44MP
availability
available only in EU
O.D. × L
26 mm × 200 mm
capacity
70 mL
thread size
25 mm , cap
wall thickness
2.0 mm
Looking for similar products? Visit Product Comparison Guide
General description
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today′s research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used.
Features and Benefits
- Manufactured from borosilicate glass 3.3
- High resistance to attack from water reduces leaching of contaminants which can cause pH changes
- PBT screwcaps with inert PTFE lined rubber disc
- Cap can withstand repeated dry heat and steam sterilization cycles.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service