Z604062
Eppendorf® Microcentrifuge 5415
Model 5415D, with rotor F 45-24-11, AC/DC input 230 V AC, 50 - 60 Hz
About This Item
Recommended Products
manufacturer/tradename
Model 5415D
availability
not available in USA and Canada
W × D × H
23 cm × 31 cm × 23 cm
Looking for similar products? Visit Product Comparison Guide
General description
Features
- Compact footprint - fits on a crowded bench
- Brushless motor; maintenance-free operation
- Spins up to 13,200 rpm; 16,110 × g
- Quick acceleration and deceleration
- Quiet operation (<60 dB)
- Adjustable-speed short-spin button-ideal for quick spins
- Accepts 2 interchangeable rotors,each with a see-through polypropylene lid
- Meets International safety standards for centrifuges (IEC 1010-2-020)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service