Key Documents
Z380156
Sigma-Aldrich
Security bag for X-ray film
For 8 × 10 inch (20 × 25 cm) film
About This Item
UNSPSC Code:
24111500
NACRES:
NB.31
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F13352-0001
General description
Contains and protects a whole box of x-ray film in a light-tight environment. The fabric is tightly woven black nylon coated inside with opaque black vinyl. The overlapping flap closure secures the bag on all edges with a hook-and-loop seal.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Xiang Li et al.
Nucleic acids research, 47(10), 5074-5085 (2019-06-05)
In microorganisms, a number of metalloproteins including PerR are found to regulate gene expression in response to environmental reactive oxygen species (ROS) changes. However, discovery of similar regulatory mechanisms remains elusive within mammalian cells. As an antioxidant metalloenzyme that maintains