Z328731
BRAND® magnetic stirring bar, PTFE, cylindrical
L 159 mm, bar diam. 27 mm, flattened sides: yes
Select a Size
1.200,00 kr.
Select a Size
About This Item
1.200,00 kr.
Recommended Products
material
PTFE-coated
cylindrical (Flattened sides)
manufacturer/tradename
BRAND 137160
L
159 mm
bar diam.
27 mm
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
- The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
- Ideal for large volumes or highly viscous mixtures.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service