Skip to Content
Merck
All Photos(2)

Key Documents

WH0051317M1

Sigma-Aldrich

Monoclonal Anti-PHF21A antibody produced in mouse

clone 5A6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BHC80, Anti-BM006, Anti-KIAA1696, Anti-PHD finger protein 21A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
3.490,00 kr.

3.490,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
3.490,00 kr.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

3.490,00 kr.


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5A6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHF21A(51317)

General description

BHC80 is a component of a BRAF35 (MIM 605535)/histone deacetylase (HDAC; see MIM 601241) complex (BHC) that mediates repression of neuron-specific genes through the cis-regulatory element known as repressor element-1 (RE1) or neural restrictive silencer (NRS) (Hakimi et al., 2002 [PubMed 12032298]).[supplied by OMIM

Immunogen

PHF21A (NP_057705, 331 a.a. ~ 430 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KQTVKSHTETDEKQTESRTITPPAAPKPKREENPQKLAFMVSLGLVTHDHLEEIQSKRQERKRRTTANPVYSGAVFEPERKKSAVTYLNSTMHPGTRKRA

Biochem/physiol Actions

The gene PHF21A (PHD finger protein 21A) is also referred to as BHC80 and forms a component of the BRAF-histone deacetylase complex, which is involved in the repression of target-gene transcription. The encoded protein is found to be essential for normal brain development and cognitive function. Disruption of this gene has been found to be associated with developmental delay and craniofacial anomalies that are seen in patients with Potocki–Shaffer syndrome (PSS).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Translocations disrupting PHF21A in the Potocki-Shaffer-syndrome region are associated with intellectual disability and craniofacial anomalies.
Kim HG
American Journal of Human Genetics, 91, 56-72 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service