Skip to Content
Merck
All Photos(1)

Key Documents

U3128

Sigma-Aldrich

Uroporphyrin I dihydrochloride

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H38N4O16 · 2HCl
CAS Number:
Molecular Weight:
903.67
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

reaction suitability

core: porphyrin
reagent type: catalyst

storage temp.

−20°C

SMILES string

Cl.Cl.OC(=O)CCc1c(CC(O)=O)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(CC(O)=O)c5CCC(O)=O)c(CC(O)=O)c4CCC(O)=O)c(CC(O)=O)c3CCC(O)=O

InChI

1S/C40H38N4O16.2ClH/c45-33(46)5-1-17-21(9-37(53)54)29-14-26-19(3-7-35(49)50)23(11-39(57)58)31(43-26)16-28-20(4-8-36(51)52)24(12-40(59)60)32(44-28)15-27-18(2-6-34(47)48)22(10-38(55)56)30(42-27)13-25(17)41-29;;/h13-16,41,44H,1-12H2,(H,45,46)(H,47,48)(H,49,50)(H,51,52)(H,53,54)(H,55,56)(H,57,58)(H,59,60);2*1H/b25-13-,26-14-,27-15-,28-16-,29-14-,30-13-,31-16-,32-15-;;

InChI key

DZLKLVAJOHLPKP-ARSPAUKVSA-N

Preparation Note

Enzymatically prepared

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S G Afonso et al.
Journal of enzyme inhibition, 10(2), 141-146 (1996-01-01)
The effect of uroporphyrin I (UI) on several cytosolic and mitochondrial enzymes (succinyl CoA synthetase, delta-aminolevulinic acid synthetase, rhodanese, lactate dehydrogenase) has been examined. All the enzymes were inactivated in the presence of the porphyrin both in the dark and
Simon Y L Ching et al.
Analytical biochemistry, 353(2), 257-265 (2006-03-28)
Few methods for assessing total antioxidant capacity (TAC) include both the percentage of inhibition and the length of inhibition in the measurement. Available methods require above ambient constant temperature incubation, reaction preheating, and/or separate assays for testing hydrophilic and hydrophobic
G Herrmann et al.
The Journal of investigative dermatology, 107(3), 398-403 (1996-09-01)
Porphyria cutanea tarda is characterized by severe connective tissue damage in sun-exposed skin. The regulated synthesis and degradation of the extracellular matrix by various matrix metalloproteinases (MMPs) determine its amount and composition within the skin. In this study, we therefore
S G Afonso et al.
Journal of enzyme inhibition, 5(3), 225-233 (1991-01-01)
The action of uroporphyrin I (URO I) on the activity of red cell uroporphyrinogen decarboxylase (URO-D) in the dark and under UV light was studied. Light-dependent-and light-independent inactivation was observed. Both effects increased at increasing concentrations of URO I, the
Remarks on the acute intermittent porphyria.
Albrecht Pfäfflin
Annals of clinical biochemistry, 46(Pt 4), 347-348 (2009-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service