Key Documents
TMO3103-0015
Nalgene® centrifuge tubes
conical bottom clear polypropylene copolymer, volume 15 mL, without cap, non-sterile, case of 100 ea
Synonym(s):
Graduated Conical Bottom Centrifuge Tubes
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
conical bottom clear polypropylene copolymer
sterility
non-sterile
feature
without cap
packaging
case of 100 ea
manufacturer/tradename
Nalgene 3103-0015
volume
15 mL
General description
Simplify measurements using Thermo Scientific Nalgene PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.
<UL><LI>Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents</LI><LI>Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (Tubes must be filled at least 80% for proper performance)</LI><LI>Conical bottoms concentrate pellet in a small area for easy isolation and retrieval</LI><LI>Molded-in graduations last the life of the tube</LI><LI>Last longer than polycarbonate tubes under conditions of repeated autoclaving</LI><LI>Tube are both autoclavable and graduated</LI></UL>
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)