Skip to Content
Merck
All Photos(1)

Key Documents

T8935

Sigma-Aldrich

BlueView Nucleic Acid Stain

in 10× TAE buffer, powder blend

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
Pricing and availability is not currently available.

grade

for molecular biology

form

powder blend

detection sensitivity measuring range

<250 ng/band

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

BlueView is a quick, safe alternative to ethidium bromide staining for nucleic acid electrophoresis. BlueView can be used directly as the running buffer and in the gel for instant staining of the bands of nucleic acids during electrophoresis, visible in ambient light. Bands can be excised from the gel without the use of UV which can cause nicking. BlueView stained bands can be used for PCR, ligation, labeling, restriction digestion, and Southern blotting.

Reconstitution

Reconstitution with water produces a 10× stock solution of the BlueView stain in 10× TAE or TBE buffer. The bottles are large enough to contain the concentrate. Dilute the 10× stock solution to the desired concentration before use. Concentrations of TBE and TAE between 0.5 and 1× are commonly used for nucleic acid electrophoresis. A 20-fold dilution of the stock solution is recommended for low background staining and optimum signal to noise.

Legal Information

BlueView is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thorunn Rafnar et al.
Nature genetics, 43(11), 1104-1107 (2011-10-04)
Ovarian cancer causes more deaths than any other gynecologic malignancy in developed countries. Sixteen million sequence variants, identified through whole-genome sequencing of 457 Icelanders, were imputed to 41,675 Icelanders genotyped using SNP chips, as well as to their relatives. Sequence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service