T4818
Tyrphostin AG 555
≥98%
Synonym(s):
N-(3′-Phenylpropyl)-3,4-dihydroxybenzylidenecyanoacetamide, Tyrphostin B46
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H18N2O3
CAS Number:
Molecular Weight:
322.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥98%
potency
IC50 (700 nM)
solubility
DMSO or ethanol: soluble
storage temp.
−20°C
SMILES string
Oc1ccc(cc1O)\C=C(/C#N)C(=O)NCCCc2ccccc2
Gene Information
human ... EGFR(1956) , ERBB2(2064)
Biochem/physiol Actions
EGFR protein tyrosine kinase inhibitor.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service