Skip to Content
Merck
All Photos(1)

Key Documents

T4271

Sigma-Aldrich

Chloranil

Synonym(s):

Tetrachloro-p-benzoquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl4O2
CAS Number:
Molecular Weight:
245.88
Beilstein:
393006
EC Number:
MDL number:
UNSPSC Code:
12352103

SMILES string

ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O

InChI

1S/C6Cl4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11

InChI key

UGNWTBMOAKPKBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service