Skip to Content
Merck
All Photos(1)

Key Documents

SRP6441

Sigma-Aldrich

DPPIV/CD26(untagged) human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

ADABP, ADCP-2, ADCP2, CD-26, DPP-4, DPP4, TP-103, TP103

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 85.4 kDa (This protein with two amino acids (Gly-Pro) at the N- terminus has a calculated MW)
observed mol wt 95 kDa by SDS-PAGE (reducing)

packaging

pkg of 10 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DPP4(1803)

General description

Dipeptidyl peptidase-IV (DPPIV), also known as adenosine deaminase complexing protein 2, DPPIV or CD26 is antigenic enzyme expressed on the surface of most cell types and is associated with immune regulation, signal transduction and apoptosis. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. The substrates of DPPIV are proline (or alanine)-containing peptides and include growth factors, chemokines, neuropeptides, and vasoactive peptides. DPPIV plays a major role in glucose metabolism. It is responsible for the degradation of incretins such as GLP-1. DPPIV plays an important role in tumor biology, and is useful as a marker for various cancers, with its levels either on the cell surface or in the serum increased in some neoplasms and decreased in others. DPPIV also binds the enzyme adenosine deaminase specifically and with high affinity. The significance of this interaction has yet to be established.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service