Skip to Content
Merck
All Photos(1)

Documents

SRP6090

Sigma-Aldrich

AKR1B10 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

AKR1B11, ALDRLn, ARL-1, Aldo-keto reductase family member B10, HIS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

36 kDa

packaging

pkg of 100 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AKR1B10(57016)

General description

AKR1B10 is a monomeric protein that efficiently catalyzes the reduction of aromatic and aliphatic aldehydes and ketones. AKR1B10 is ubiquitously expressed in many human tissues but is highly expressed in small intestine, colon and adrenal gland. This protein is pathogenically involved in diabetic complications. It has been reported that AKR1B10 is overexpressed in human tumors, such as liver, breast, and lung cancer, and may play a critical role in the development and progression of cancer.

Physical form

1 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0) containing 10% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MATFVELSTK AKMPIVGLGT WKSPLGKVKE AVKVAIDAGY RHIDCAYVYQ NEHEVGEAIQ EKIQEKAVKR EDLFIVSKLW PTFFERPLVR KAFEKTLKDL KLSYLDVYLI HWPQGFKSGD DLFPKDDKGN AIGGKATFLD AWEAMEELVD EGLVKALGVS NFSHFQIEKL LNKPGLKYKP VTNQVECHPY LTQEKLIQYC HSKGITVTAY SPLGSPDRPW AKPEDPSLLE DPKIKEIAAK HKKTAAQVLI RFHIQRNVIV IPKSVTPARI VENIQVFDFK LSDEEMATIL SFNRNWRACN VLQSSHLEDY PFDAEY

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yihao Tian et al.
PloS one, 6(2), e17017-e17017 (2011-03-03)
Oncogenes and tumor suppressors work in concert to regulate cell growth or death, which is a pair of antagonist factors for regulation of tumorigenesis. Here we show promoter characteristic of tumor suppressor RASSF1A, which revealed a p53 binding site in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service