Skip to Content
Merck
All Photos(1)

Key Documents

SRP6058

Sigma-Aldrich

Kallikrein-3 human

recombinant, expressed in human cells, ≥95% (SDS-PAGE)

Synonym(s):

APS, Hk3, KLK2A1, Kallikrein-3, P-30 antigen, PSA

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
4.210,00 kr.

4.210,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
4.210,00 kr.

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

4.210,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in human cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

35 kDa

packaging

pkg of 20 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KLK3(354)

Related Categories

General description

Prostate-specific antigen, also known as Gamma-seminoprotein, Kallikrein-3, kallikrein-related peptidase 3, Semenogelase, KLK3 and PSA, is a secreted protein which belongs to thepeptidase S1 family and Kallikrein subfamily. KLK3/Kallikrein 3 contains one peptidase S1 domain. KLK3/Kallikrein 3 is a glycoprotein produced almost exclusively by the prostate gland. KLK3/Kallikrein 3 is produced for the ejaculate where it liquifies the semen in the seminal coagulum and allows sperm to swim freely.It is also believed to be instrumental in dissolving the cervical mucous, allowing the entry of sperm. Human KLK3/Kallikrein 3 and human KLK2 are closely related products of the human kallikrein genes KLK3 and KLK2, respectively. Both KLK3/Kallikrein 3 and human kallikrein 2 are produced and secreted in the prostate and have important applications in the diagnosis of prostate cancer. Understanding the mechanism by which genetic variation in KLK3/Kallikrein 3 affects prostate cancer risk has important implications for study of the biological role of KLK3/Kallikrein 3 in prostate tumorigenesis.

Physical form

Lyophilized from sterile PBS, pH 7.4. (Normally 5 % - 8 % trehalose and mannitol are added as protectants before lyophilization.)

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Detailed reconstitution instructions are sent along with the product.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service