Skip to Content
Merck
All Photos(2)

Key Documents

SRP5338

Sigma-Aldrich

PP1A, active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

LAR

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
2.930,00 kr.

2.930,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
2.930,00 kr.

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

2.930,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

182-274 nmol/min·mg

mol wt

~62 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PPP1CA(5499)

General description

PP1A is a serine/threonine specific protein phosphatase family which is involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. PP1A is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1A functions as a suppressor of learning and memory and as a potential mediator of cognitive decline during aging.PP1A is also play a main role in splice site selection and is an important regulator of cardiac function.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service